Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAS40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531178 Supplier Invitrogen™ Supplier No. PA531178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12. Predicted reactivity: Mouse (94%), Rat (92%), Chimpanzee (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The Akt signaling pathway contributes to the regulation of apoptosis after a variety of cell death signals. AKT1S1, also known as PRAS40, is a proline-rich substrate of the kinase AKT1 and is thought to play a role in neuroprotection mediated by nerve growth factor (NGF) after transient focal cerebral ischemia. AKT1S1 is also a substrate and potential regulator of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Treatment with the insulin-like growth factor-1 (IGF1) can induce the phosphorylation of AKT1S1 via the PI3K/AKT signaling pathway in PC12 cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAS40
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene AKT1S1
Gene Accession No. Q96B36, Q9D1F4
Gene Alias 1110012J22Rik; 40 kDa proline-rich AKT substrate; AI227026; AKT1 substrate 1; AKT1 substrate 1 (proline rich); AKT1 substrate 1 (proline-rich); akt1s1; Lobe; Lobel; OTTHUMP00000196756; OTTHUMP00000196760; PKB/Akt substrate 40; Pras; PRAS40; Proline-rich AKT substrate; proline-rich AKT1 substrate 1
Gene Symbols AKT1S1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PRAS40. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292887, 67605, 84335
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.