Learn More
Invitrogen™ PRAS40 Recombinant Rabbit Monoclonal Antibody (2Y8Y3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542806
Description
Positive test controls include: HeLa, MCF7, NIH/3T3, RD. The target is usually found in the following locations: Cytoplasm, cytosol. Immunogen sequence: STDDGSLSEE TPAGPPTCSV PPASALPTQQ YAKSLPVSVP VWGFKEKRTE ARSSDEENGP PSSPDLDRIA ASMRALVLRE AEDTQVFGDL PRPRLNTSDF QKLKRKY.
The Akt signaling pathway contributes to the regulation of apoptosis after a variety of cell death signals. AKT1S1, also known as PRAS40, is a proline-rich substrate of the kinase AKT1 and is thought to play a role in neuroprotection mediated by nerve growth factor (NGF) after transient focal cerebral ischemia. AKT1S1 is also a substrate and potential regulator of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Treatment with the insulin-like growth factor-1 (IGF1) can induce the phosphorylation of AKT1S1 via the PI3K/AKT signaling pathway in PC12 cells.
Specifications
PRAS40 | |
Recombinant Monoclonal | |
1.05 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96B36, Q9D1F4 | |
AKT1S1 | |
A synthetic peptide corresponding to a sequence within amino acids 150-256 of human AKT1S1/PRAS40 (Q96B36). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
2Y8Y3 | |
Unconjugated | |
AKT1S1 | |
1110012J22Rik; 40 kDa proline-rich AKT substrate; AI227026; AKT1 substrate 1; AKT1 substrate 1 (proline rich); AKT1 substrate 1 (proline-rich); akt1s1; Lobe; Lobel; OTTHUMP00000196756; OTTHUMP00000196760; PKB/Akt substrate 40; Pras; PRAS40; Proline-rich AKT substrate; proline-rich AKT1 substrate 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
67605, 84335 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.