Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium(III) nitrate hexahydrate, REacton™, 99.99% (REO)

Catalog No. aa1124014
Click to view available options
Quantity:
25 g
100 g

CAS: 15878-77-0 | H12N3O15Pr | 435.01 g/mol

Praseodymium(III) nitrate hexahydrate is used in electrical components, optical glasses and photo-optical materials. Also find use as catalysts and laboratory reagent. In addition, it is used in the solvothermal process for formation of doped lanthanide oxysulfide. It is a precursor for the electrodeposition of praseodymium oxide for electrochemical applications. It is a starting material for the preparation of ceramic pigments, metal-organic frameworks and mixed conductors. It finds utility in the synthesis of other praseodymium compounds and complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15878-77-0
Molecular Formula H12N3O15Pr
Molecular Weight (g/mol) 435.01
MDL Number MFCD00149826
InChI Key LXXCECZPOWZKLC-UHFFFAOYSA-N
Synonym praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrate
PubChem CID 204170
IUPAC Name praseodymium(3+);trinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 25 g
UN Number UN1477
Solubility Information Soluble in water and alcohol.
Formula Weight 435.01 (326.92 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Praseodymium(III) nitrate hexahydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 233-796-5
RTECSNumber TU1400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.