Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium(III) nitrate hydrate, 99.9% (REO)

Catalog No. AA12909A1
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg

CAS: 15878-77-0 | H12N3O15Pr | 435.01 g/mol

Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15878-77-0
Molecular Formula H12N3O15Pr
Molecular Weight (g/mol) 435.01
MDL Number MFCD00011175
InChI Key LXXCECZPOWZKLC-UHFFFAOYSA-N
Synonym praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrate
PubChem CID 204170
SMILES O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Green
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99.9%
Solubility Information Soluble in water,anhydrous amine,alcohol,ether,polar solvents and acetonitrile.
IUPAC Name praseodymium(3+);trinitrate;hexahydrate
Formula Weight 326.92 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Green crystals
Chemical Name or Material Praseodymium(III) nitrate hydrate, Rare Earth Oxides Content
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 233-796-5
RTECSNumber TU1400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.