Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium(III) nitrate pentahydrate, 99.9%, (trace metal basis)

Catalog No. ac194590500
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle

CAS: 14483-17-1 | N3O9Pr·5H2O | 417 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14483-17-1
Molecular Formula N3O9Pr·5H2O
Molecular Weight (g/mol) 417
MDL Number MFCD00149825

Specifications

Color Green
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99.90%
Linear Formula Pr(NO3)3·5H2O
Formula Weight 417
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Praseodymium(III) nitrate pentahydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.