Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Praseodymium(III) nitrate hexahydrate

Catalog No. 205133250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g
50 g
250 g

Catalog No. 20-513-3250G

Supplier: Sigma Aldrich 205133250G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A novel solvothermal process for formation of doped lanthanide oxysulfide, La2O2S, was developed in order to advance possible applications of this material in phosphors, fluorescent display tubes or heat transfer measurement. Also applied in the ultrasonic synthesis of phase-pure praseodymium molybate,Pr2Mo3O12.

Specifications

CAS 15878-77-0
Molecular Formula N3O9Pr -+ 6H2O
Linear Formula Pr(NO3)3-+6H2O
Quantity 250 g
MDL Number MFCD00149826
Synonym Praseodymium trinitrate hexahydrate
Molecular Weight (g/mol) 435.01
Percent Purity 99.9% trace metals basis
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.