Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prazosin hydrochloride

Catalog No. aaj61712md
Click to view available options
Quantity:
250 mg
1 g

An alpha adrenoceptor antagonist | CAS: 19237-84-4 | C19H22ClN5O4 | 419.87 g/mol

Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin

Solubility
Soluble in dimethyl sulfoxide, ethanol, water, methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19237-84-4
Molecular Formula C19H22ClN5O4
Molecular Weight (g/mol) 419.87
MDL Number MFCD00058177
InChI Key WFXFYZULCQKPIP-UHFFFAOYSA-N
Synonym prazosin hydrochloride, prazosin hcl, minipress, vasoflex, peripress, furazosin hydrochloride, deprazolin, hypovase, hypovasole, pratsiol
PubChem CID 68546
ChEBI CHEBI:8365
IUPAC Name hydrogen 2-[4-(furan-2-carbonyl)piperazin-1-yl]-6,7-dimethoxyquinazolin-4-amine chloride
SMILES [H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1=CC=CO1

Specifications

Melting Point 285°C
Color White
Quantity 250 mg
Beilstein 4303561
Merck Index 14,7717
Solubility Information Soluble in dimethyl sulfoxide,ethanol,water,methanol.
Formula Weight 419.86
Physical Form Powder
Chemical Name or Material Prazosin hydrochloride
Hazard Category H302-H315-H319-H335-H361
Hazard Statement GHS H Statement
H361-H302-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 242-903-4
RTECSNumber VA1350000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.