Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598084
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598084 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598084 Supplier Invitrogen™ Supplier No. PA598084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRB1 Polyclonal Antibody for Western Blot, ELISA

This gene encodes a member of the heterogeneous family of basic, proline-rich, human salivary glycoproteins. The encoded preproprotein undergoes proteolytic processing to generate one or more mature peptides before secretion from the parotid glands. Multiple alleles of this gene exhibiting variations in the length of the tandem repeats have been identified. The reference genome encodes the Medium allele. This gene is located in a cluster of closely related salivary proline-rich proteins on chromosome 12. Alternative splicing results in multiple transcript variants encoding different isoforms that may undergo similar proteolytic processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRB1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PRB1
Gene Accession No. P04280
Gene Alias Basic peptide IB-6; Basic salivary proline-rich protein 1; BstNI type basic salivary proline-rich protein 1; parotid middle band protein; Peptide P-H; PM; PMF; PMS; PRB1; PRB1L; PRB1M; proline rich protein BstNI subfamily 1; Proline-rich peptide II-2; proline-rich protein BstNI subfamily 1; Salivary proline-rich protein; salivary protein Pe
Gene Symbols PRB1
Host Species Rabbit
Immunogen Recombinant Human Basic salivary proline-rich protein 1 protein (17-92AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5542
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.