Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pRb2 p130 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991980
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1980 100 μg
50-199-1981 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1980 Supplier Rockland Immunochemicals Supplier No. 600401B11
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antiserum is directed against Spa310 and reacts with the Spa310 domain of pRb2/p130 from human tissues. Based on the sequence we expect this antibody to react as well Spa310 from chimpanzee and orangutan, and with lesser affinity, Spa310 from horse, dog, bovine, rat and opossum.

RBL2 (also known as p130) or Retinoblastoma-like protein 2 is a key regulator of entry into cell division. It is directly involved in heterochromatin formation by maintaining overall chromatin structure and (in particular) that of constitutive heterochromatin by stabilizing histone methylation. RBL2 recruits and targets histone methyltransferases KMT5B and KMT5C leading to epigenetic transcriptional repression. It controls histone H4 'Lys20' trimethylation. It probably also acts as a transcription repressor by recruiting chromatin-modifying enzymes to promoters. RBL2 serves as a potent inhibitor of E2F-mediated trans-activation and associates preferentially with E2F5.

Specifications

Antigen pRb2 p130
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.39 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.1% sodium azide; pH 7.2
Gene RBL2
Gene Accession No. Q08999
Gene Alias 130 kDa retinoblastoma-associated protein; FLJ26459; p130; PPAR-alpha-interacting complex protein 128; pRb2; PRIC128; RB transcriptional corepressor like 2; RB2; Rbl2; RBR-2; retinoblastoma-like 2; Retinoblastoma-like protein 2; Retinoblastoma-related protein 2
Gene Symbols RBL2
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 467978, 5934
Target Species Chimpanzee, Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.