Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRC1 Monoclonal Antibody (16F2)
GREENER_CHOICE

Catalog No. PIMA1846
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1846 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1846 Supplier Invitrogen™ Supplier No. MA1846
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-846 detects PRC1 from human, mouse, and rat samples. MA1-846 has been successfully used in Western blot, immunoprecipitation, and immunofluorescence procedures. The MA1-846 immunogen is recombinant PRC1 protein.

This gene encodes a protein that is involved in cytokinesis. The protein is present at high levels during the S and G2/M phases of mitosis but its levels drop dramatically when the cell exits mitosis and enters the G1 phase. It is located in the nucleus during interphase, becomes associated with mitotic spindles in a highly dynamic manner during mitosis, and localizes to the cell mid-body during cytokinesis. This protein has been shown to be a substrate of several cyclin-dependent kinases (CDKs). It is necessary for polarizing parallel microtubules and concentrating the factors responsible for contractile ring assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRC1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 16F2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene PRC1
Gene Accession No. O43663, Q99K43
Gene Alias anaphase spindle elongation 1 homolog; ASE1; carboxypeptidase C PRC1; Carboxypeptidase Y; Carboxypeptidase YSCY; CPD-Y; CPY1; D7Ertd348e; LBC1; MGC1671; MGC3669; prc1; protein regulating cytokinesis 1; protein regulator of cytokinesis 1; YMR297W
Gene Symbols PRC1
Host Species Mouse
Immunogen Recombinant PRC1 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233406, 308761, 9055
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.