Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596553
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596553 Supplier Invitrogen™ Supplier No. PA596553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LFEGVQKWEE TWRLFLEFER KASDPNRFTN RGGNLLKEEK QRAKLQKMLP KLEEELKARI ELWEQEHSKA FMVNGQKFME YVAEQWEMHR LEKERAKQER QLKNKKQTET EMLYGSAPRT PSKRRGLAPN TPGKARKLNT TTMSNATANS SIRPIFGGTV YHSPVSRLPP SGSKPVAAST CSGKKTPRTG RHGANKENLE LNGSILSGGY PGSAPLQRNF SINSVASTYS EFAKDPSLSD SSTVGLQREL SKASKSDATS GILNSTNIQS; Positive Samples: MCF7, Jurkat, 293T, HepG2; Cellular Location: Cytoplasm, Midbody, Nucleus, cytoskeleton, spindle pole.

This gene encodes a protein that is involved in cytokinesis. The protein is present at high levels during the S and G2/M phases of mitosis but its levels drop dramatically when the cell exits mitosis and enters the G1 phase. It is located in the nucleus during interphase, becomes associated with mitotic spindles in a highly dynamic manner during mitosis, and localizes to the cell mid-body during cytokinesis. This protein has been shown to be a substrate of several cyclin-dependent kinases (CDKs). It is necessary for polarizing parallel microtubules and concentrating the factors responsible for contractile ring assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRC1
Gene Accession No. O43663, Q99K43
Gene Alias anaphase spindle elongation 1 homolog; ASE1; carboxypeptidase C PRC1; Carboxypeptidase Y; Carboxypeptidase YSCY; CPD-Y; CPY1; D7Ertd348e; LBC1; MGC1671; MGC3669; prc1; protein regulating cytokinesis 1; protein regulator of cytokinesis 1; YMR297W
Gene Symbols PRC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 351-620 of human PRC1 (NP_0039721).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233406, 308761, 9055
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.