Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRCC Monoclonal Antibody (OTI5H7), TrueMAB™, OriGene
SDP

Catalog No. 501684702
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4702 Supplier OriGene Supplier No. TA812966
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRCC may regulate cell cycle progression through interaction with MAD2L2.

Specifications

Antigen PRCC
Applications Western Blot
Classification Monoclonal
Clone OTI5H7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PRCC
Gene Accession No. Q92733
Gene Alias RCCP1, TPRC
Gene Symbols Prcc
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 135-491 of human PRCC produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5546, 94315
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.