Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRCC Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553993
Description
Immunogen sequence: RNRGREEINF VEIKGDDQLS GAQQWMTKSL TEEKTMKSFS KKKGEQPTGQ QRRKHQITYL IHQAKERELE LKNTWSENKL SRRQTQAKY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
PRCC may regulate cell cycle progression through interaction with MAD2L2.
Specifications
PRCC | |
Polyclonal | |
Unconjugated | |
Prcc | |
LOW QUALITY PROTEIN: proline-rich protein PRCC; papillary renal cell carcinoma (translocation-associated); papillary renal cell carcinoma translocation-associated gene product; papillary renal cell carcinoma translocation-associated gene protein; PRCC; PRCC, proline rich mitotic checkpoint control factor; proline rich mitotic checkpoint control factor; proline-rich protein PRCC; RCCP1; si:ch211-210h11.8; TPRC; Unknown (protein for MGC:140094) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5546 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92733 | |
Prcc | |
Recombinant protein corresponding to Human PRCC. Recombinant protein control fragment (Product #RP-92570). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction