Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRCC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553998 Supplier Invitrogen™ Supplier No. PA553998
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDEAEPEPEE EEAVAPTSGP ALGGLFASLP APKGPALLPP PPQMLAPAFP PPLLLPPPTG DPRLQPPPPL PFGLGGFPPP PGVSPAEAAG VGEGLGLGLP S Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

PRCC may regulate cell cycle progression through interaction with MAD2L2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRCC
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Prcc
Gene Accession No. Q92733
Gene Alias LOW QUALITY PROTEIN: proline-rich protein PRCC; papillary renal cell carcinoma (translocation-associated); papillary renal cell carcinoma translocation-associated gene product; papillary renal cell carcinoma translocation-associated gene protein; PRCC; PRCC, proline rich mitotic checkpoint control factor; proline rich mitotic checkpoint control factor; proline-rich protein PRCC; RCCP1; si:ch211-210h11.8; TPRC; Unknown (protein for MGC:140094)
Gene Symbols Prcc
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRCC. Recombinant protein control fragment (Product #RP-92571).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5546
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.