Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRCC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570356 Supplier Invitrogen™ Supplier No. PA570356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

PRCC may regulate cell cycle progression through interaction with MAD2L2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRCC
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Prcc
Gene Accession No. 0
Gene Alias LOW QUALITY PROTEIN: proline-rich protein PRCC; papillary renal cell carcinoma (translocation-associated); papillary renal cell carcinoma translocation-associated gene product; papillary renal cell carcinoma translocation-associated gene protein; PRCC; PRCC, proline rich mitotic checkpoint control factor; proline rich mitotic checkpoint control factor; proline-rich protein PRCC; RCCP1; si:ch211-210h11.8; TPRC; Unknown (protein for MGC:140094)
Gene Symbols Prcc
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat PRCC.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310687
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.