Learn More
PRCC Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302340AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
PRCC may regulate cell cycle progression through interaction with MAD2L2.Specifications
PRCC | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q92733 | |
Prcc | |
Between 425 and 475 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PRCC | |
papillary renal cell carcinoma translocation-associated gene protein, RCCP1, TPRC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5546, 94315 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.