Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRCC Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PRCC |
---|---|
Concentration | 0.24 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7647
![]() ![]() |
Proteintech Group Inc
120191AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
PRCC may regulate cell cycle progression through interaction with MAD2L2.Specifications
PRCC | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q92733 | |
5546 | |
PRCC Fusion Protein Ag2638 | |
Primary | |
-20°C | |
PRCC |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PRCC, Proline rich protein PRCC, RCCP1, TPRC | |
PRCC | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title