Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRCP (Angiotensinase C) Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277553
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7553 Supplier Leinco Technologies Inc Supplier No. 13920100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRCP (Angiotensinase C) Polyclonal antibody specifically detects PRCP (Angiotensinase C) in Human samples. It is validated for ELISA

Specifications

Antigen PRCP (Angiotensinase C)
Applications ELISA
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% sodium azide
Gene PRCP
Gene Accession No. P42785
Gene Alias 2510048K03Rik; 2610104A14Rik; AI451719; Angiotensinase C; HUMPCP; Lysosomal carboxypeptidase C; lysosomal Pro-X carboxypeptidase; PCP; Prcp; Proline carboxypeptidase; prolylcarboxypeptidase; prolylcarboxypeptidase (angiotensinase C)
Host Species Rabbit
Immunogen Recombinant fusion protein corresponding to aa 46-317 of human PRCP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5547
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.