Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRCP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119360
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119360 200 μL
PIPA5121671 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119360 Supplier Invitrogen™ Supplier No. PA5119360
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRCP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Cleaves C-terminal amino acids linked to proline in peptides such as angiotensin II, III and des-Arg9-bradykinin. This cleavage occurs at acidic pH, but enzymatic activity is retained with some substrates at neutral pH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRCP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PRCP
Gene Accession No. Q7TMR0
Gene Alias 2510048K03Rik; 2610104A14Rik; AI451719; Angiotensinase C; HUMPCP; Lysosomal carboxypeptidase C; lysosomal Pro-X carboxypeptidase; PCP; Prcp; Proline carboxypeptidase; prolylcarboxypeptidase; prolylcarboxypeptidase (angiotensinase C)
Gene Symbols PRCP
Host Species Rabbit
Immunogen Recombinant protein Prolylcarboxypeptidase. The antigen corresponds to amino acid range 169-443 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72461
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.