Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRCP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553543 Supplier Invitrogen™ Supplier No. PA553543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KHLKRTIPGA ENQPVIAIGG SYGGMLAAWF RMKYPHMVVG ALAASAPIWQ FEDLVPCGVF MKIVTTDFRK SGPHCSESIH RSWDAINRLS NTGSGLQWLT GALHLCSPLT SQDIQHLKDW ISETWVNLAM VDYP Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 78%.

Cleaves C-terminal amino acids linked to proline in peptides such as angiotensin II, III and des-Arg9-bradykinin. This cleavage occurs at acidic pH, but enzymatic activity is retained with some substrates at neutral pH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRCP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRCP
Gene Accession No. P42785
Gene Alias 2510048K03Rik; 2610104A14Rik; AI451719; Angiotensinase C; HUMPCP; Lysosomal carboxypeptidase C; lysosomal Pro-X carboxypeptidase; PCP; Prcp; Proline carboxypeptidase; prolylcarboxypeptidase; prolylcarboxypeptidase (angiotensinase C)
Gene Symbols PRCP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRCP. Recombinant protein control fragment (Product #RP-90271).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5547
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.