Learn More
Invitrogen™ PRCP Polyclonal Antibody

Description
Immunogen sequence: ITTMYGGKNI SSHTNIVFSN GELDPWSGGG VTKDITDTLV AVTISEGAHH LDLRTKNALD PMSVLLARSL EVRHMKNWIR DFYDSAGKQH.
Specifications
Specifications
| Antigen | PRCP |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.19 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PRCP |
| Gene Accession No. | 0 |
| Gene Alias | 2510048K03Rik; 2610104A14Rik; AI451719; Angiotensinase C; HUMPCP; Lysosomal carboxypeptidase C; lysosomal Pro-X carboxypeptidase; PCP; Prcp; Proline carboxypeptidase; prolylcarboxypeptidase; prolylcarboxypeptidase (angiotensinase C) |
| Gene Symbols | PRCP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.