Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM1 (BLIMP-1) Mouse anti-Human, Unconjugated, Clone: ROS195G also called 195G/G5, MilliporeSigma™

Catalog No. MABE18141MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE18141MI 100 μg
MABE18142MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE18141MI Supplier MilliporeSigma Supplier No. MABE1814100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRDM1 (BLIMP-1) Monoclonal antibody specifically detects PRDM1 (BLIMP-1) in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM1 (BLIMP-1)
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone ROS195G also called 195G/G5
Conjugate Unconjugated
Gene Accession No. NP_878911.1
Gene Alias PR domain zinc finger protein 1;BLIMP-1;Beta-interferon gene positive regulatory domain I-binding factor;PR domain-containing protein 1;Positive regulatory domain I-binding factor 1;PRDI-BF1;PRDI-binding factor 1
Gene Symbols PRDM1, BLIMP1
Host Species Mouse
Immunogen GST-tagged full-length human recombinant PR domain zinc finger protein 1.
Molecular Weight of Antigen 91.77 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone ROS195G is a mouse monoclonal antibody that detects PR domain zinc finger protein 1.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.