Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582948 Supplier Invitrogen™ Supplier No. PA582948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPSHIQGSSS TQGQALQQQQ QQQQNSSVQH TYLPSAWNSF RGYSSEIQMM TLPPGQFVIT DSGVATPVTT GQVKAVTSGH YVLSESQSEL EEKQTSALSG GVQVEPPAHS DSLDPQTNSQ QQTTQYIITT TTNGNGS Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 88%.

GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM10
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRDM10
Gene Accession No. Q9NQV6
Gene Alias Gm1112; KIAA1231; MGC131802; PFM7; PR domain 10; PR domain containing 10; PR domain zinc finger protein 10; PR domain-containing protein 10; PRDM zinc finger transcription factor; PRDM10; PR-domain family member 7; RGD1560384; TRIS; tristanin
Gene Symbols PRDM10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRDM10. Recombinant protein control fragment (Product #RP-91659).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56980
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.