Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597091 Supplier Invitrogen™ Supplier No. PA597091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DYMKRLHSMS QETIHRNLAR GEKRLQREKS EQVLDNPEDL RGPIHLSVLR QGKSPYKRGF DEGDVHPQAK KKKIDLIFKD VLEASLESAK VEAHQLALST SLVIRKVPKY QDDAYSQCAT TMTHGVQNIG QTQGEGDWKV PQGVSKEPGQ LEDEEEEPSS FKADSPAEAS LASDPHELPT TSFCPNCIRL KKKVRELQAE LDMLKSGKLP EPPVLPPQVL ELPEFSDPAG KLVWMRLLSE GRVRSGLCGG; Positive Samples: Mouse heart, Mouse lung; Cellular Location: Cytoplasm, Nucleus.

PRDM11 (PR/SET Domain 11) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and methyltransferase activity. May be involved in transcription regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRDM11
Gene Accession No. A2AGX3
Gene Alias 8030443D09Rik; AA163972; Gm707; PFM8; PR domain 11; PR domain containing 11; PR domain-containing protein 11; PRDM11; PR-domain containing protein 11; RGD1305673
Gene Symbols PRDM11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 228-477 of human PRDM11 (NP_0012436251).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100042784
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.