Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRDM11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598964
Description
PRDM11 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
PRDM11 (PR/SET Domain 11) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and methyltransferase activity. May be involved in transcription regulation.
Specifications
PRDM11 | |
Polyclonal | |
Unconjugated | |
PRDM11 | |
8030443D09Rik; AA163972; Gm707; PFM8; PR domain 11; PR domain containing 11; PR domain-containing protein 11; PRDM11; PR-domain containing protein 11; RGD1305673 | |
Rabbit | |
Protein G | |
RUO | |
56981 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2.78 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9NQV5 | |
PRDM11 | |
Recombinant Human PR domain-containing protein 11 protein (63-215AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction