Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560257
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560257 Supplier Invitrogen™ Supplier No. PA560257
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MWEVFNEDGT VRYFIDASQE DHRSWMTYIK CARNEQEQNL EVVQIGTSIF YKAIEMIPPD QELLVWYGNS HNTF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a transcriptional regulator of sensory neuronal specification that plays a critical role in pain perception. The encoded protein contains an N-terminal PRDI-BF1 and RIZ homology (PR) domain, a SET domain, and three C-terminal C2H2 zinc finger DNA-binding domains. Naturally occurring mutations in this gene are associated with congenital insensitivity to pain (CIP), and hereditary sensory and autonomic neuropathies (HSAN's) affecting peripheral sensory and autonomic neurons. Deregulation of this gene is associated with solid cancers and hematological malignancies including chronic myeloid leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM12
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRDM12
Gene Accession No. A2AJ77, Q9H4Q4
Gene Alias Gm998; HSAN8; PFM9; PR domain 12; PR domain containing 12; PR domain zinc finger protein 12; PR domain-containing protein 12; PRDM12; PR-domain containing protein 12; PR-domain zinc finger protein 12; RP11-57C19.7
Gene Symbols PRDM12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRDM12. Recombinant protein control fragment (Product #RP-98426).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381359, 59335
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.