Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRDM12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101259
Description
Antibody detects endogenous levels of total PRDM12.
This gene encodes a transcriptional regulator of sensory neuronal specification that plays a critical role in pain perception. The encoded protein contains an N-terminal PRDI-BF1 and RIZ homology (PR) domain, a SET domain, and three C-terminal C2H2 zinc finger DNA-binding domains. Naturally occurring mutations in this gene are associated with congenital insensitivity to pain (CIP), and hereditary sensory and autonomic neuropathies (HSAN's) affecting peripheral sensory and autonomic neurons. Deregulation of this gene is associated with solid cancers and hematological malignancies including chronic myeloid leukemia.
Specifications
PRDM12 | |
Polyclonal | |
Unconjugated | |
PRDM12 | |
Gm998; HSAN8; PFM9; PR domain 12; PR domain containing 12; PR domain zinc finger protein 12; PR domain-containing protein 12; PRDM12; PR-domain containing protein 12; PR-domain zinc finger protein 12; RP11-57C19.7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
381359, 59335 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A2AJ77, Q9H4Q4 | |
PRDM12 | |
A synthesized peptide derived from human PRDM12(Accession Q9H4Q4), corresponding to amino acid residues L198-C248. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction