Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRDM13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | PRDM13 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18036020
![]() |
Novus Biologicals
NBP18036020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180360
![]() |
Novus Biologicals
NBP180360 |
100 μL |
Each for $487.50
|
|
|||||
Description
PRDM13 Polyclonal specifically detects PRDM13 in Human samples. It is validated for Western Blot.Specifications
| PRDM13 | |
| Polyclonal | |
| Rabbit | |
| NP_067633 | |
| 59336 | |
| Synthetic peptide directed towards the N terminal of human PRDM13. Peptide sequence HGAARAPATSVSADCCIPAGLRLGPVPGTFKLGKYLSDRREPGPKKKVRM. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PR domain containing 13, PR domain zinc finger protein 13 | |
| PRDM13 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title