Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113483
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113483 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113483 Supplier Invitrogen™ Supplier No. PA5113483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRDM13 Polyclonal Antibody for Western Blot, ELISA

PRDM13 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM13
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PRDM13
Gene Accession No. E9PZZ1, Q9H4Q3
Gene Alias MU-MB-20.220; PFM10; PR domain 13; PR domain containing 13; PR domain zinc finger protein 13; PR domain-containing protein 13; PR/SET domain 13; PRDM13; PR-domain containing protein 13; PR-domain zinc finger protein 13
Gene Symbols PRDM13
Host Species Rabbit
Immunogen Recombinant Human PR domain zinc finger protein 13 protein (2-255AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230025, 59336
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.