Learn More
Invitrogen™ PRDM14 Monoclonal Antibody (GT1359)

Description
Predicted Reactivity: Mouse (82%), Bovine (91%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PRDM14 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT1359 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Prdm14 |
| Gene Accession No. | Q9GZV8 |
| Gene Alias | LOC100365184; PFM11; PR domain 14; PR domain containing 14; PR domain containing 14-like; PR domain zinc finger protein 14; PR domain-containing 14; PR domain-containing protein 14; PR/SET domain 14; PRDM14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.