Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM14 Mouse anti-Human, Rat, Unconjugated, Clone: 4F10, Abnova™

Catalog No. 89335129 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-129 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-129 Supplier Abnova Corporation Supplier No. H00063978M01
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

PRDM14 Monoclonal antibody specifically detects PRDM14 in Human, Rat samples. It is validated for WB-Ce, WB-Ti, ELISA.

Specifications

Antigen PRDM14
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4F10
Conjugate Unconjugated
Description PR domain containing 14
Gene PRDM14
Gene Accession No. BC052311.1
Gene Alias MGC59730, PFM11
Host Species Mouse
Immunogen A synthetic peptide corresponding to human PRDM14
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 63978
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.