Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRDM14 Mouse anti-Human, Rat, Unconjugated, Clone: 5H2, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation H00063978M02
Description
PRDM14 Monoclonal antibody specifically detects PRDM14 in Human, Rat samples. It is validated for WB-Ce, WB-Ti, ELISA.Specifications
PRDM14 | |
Monoclonal | |
Unconjugated | |
PRDM14 | |
MGC59730, PFM11 | |
A synthetic peptide corresponding to human PRDM14 | |
Primary | |
Human, Rat | |
IgG2b κ |
Western Blot, ELISA | |
5H2 | |
PR domain containing 14 | |
BC052311.1 | |
Mouse | |
100 μg | |
63978 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction