Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540682 Supplier Invitrogen™ Supplier No. PA540682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GVTPSLEHPA SLHHAISGLL VPPDSSGSDS LPQTLDKDSL QLPEGLCLMQ Sequence homology: Cow: 86%; Dog: 86%; Horse: 86%; Human: 100%; Pig: 86%; Rabbit: 100%.

PRDM14 encodes a member of the PRDI-BF1 and RIZ homology domain containing (PRDM) family of transcriptional regulators. The encoded protein may possess histone methyltransferase activity and plays a critical role in cell pluripotency by suppressing the expression of differentiation marker genes. Expression of this gene may play a role in breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM14
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Prdm14
Gene Accession No. Q9GZV8
Gene Alias LOC100365184; PFM11; PR domain 14; PR domain containing 14; PR domain containing 14-like; PR domain zinc finger protein 14; PR domain-containing 14; PR domain-containing protein 14; PR/SET domain 14; PRDM14
Gene Symbols Prdm14
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PRDM14.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63978
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.