Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRDM14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593019
Description
Positive Samples: Mouse liver; Cellular Location: Nucleus.
PRDM14 encodes a member of the PRDI-BF1 and RIZ homology domain containing (PRDM) family of transcriptional regulators. The encoded protein may possess histone methyltransferase activity and plays a critical role in cell pluripotency by suppressing the expression of differentiation marker genes. Expression of this gene may play a role in breast cancer.
Specifications
PRDM14 | |
Polyclonal | |
Unconjugated | |
Prdm14 | |
LOC100365184; PFM11; PR domain 14; PR domain containing 14; PR domain containing 14-like; PR domain zinc finger protein 14; PR domain-containing 14; PR domain-containing protein 14; PR/SET domain 14; PRDM14 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100365184, 383491, 63978 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E9Q3T6, Q9GZV8 | |
Prdm14 | |
Recombinant protein of mouse PRDM14. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction