Learn More
Invitrogen™ PRDM14 Polyclonal Antibody

Description
PRDM14 Polyclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | PRDM14 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide; pH 7.2-7.4 |
| Gene | Prdm14 |
| Gene Accession No. | Q9GZV8 |
| Gene Alias | LOC100365184; PFM11; PR domain 14; PR domain containing 14; PR domain containing 14-like; PR domain zinc finger protein 14; PR domain-containing 14; PR domain-containing protein 14; PR/SET domain 14; PRDM14 |
| Gene Symbols | Prdm14 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.