Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRDM15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541210
Description
Peptide sequence: SILTVTFDTV SGSAMLHNRQ NDVQIHPQPE ASNPQSVAHF INLTTLVNSI Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 79%; Human: 100%; Mouse: 100%; Pig: 93%; Rat: 100%.
The exact function of PRDM15 remains unknown.
Specifications
PRDM15 | |
Polyclonal | |
Unconjugated | |
PRDM15 | |
C21orf83; E130018M06Rik; ORF62; PFM12; PFM15; PR domain 15; PR domain containing 15; PR domain zinc finger protein 15; PR domain-containing protein 15; PR/SET domain 15; PRDM15; Zfp298; Zinc finger protein 298; ZNF298 | |
Rabbit | |
Affinity chromatography | |
RUO | |
63977 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P57071 | |
PRDM15 | |
Synthetic peptide directed towards the C-terminal of human PRDM15 (aa 1405-1454). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction