Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565041 Supplier Invitrogen™ Supplier No. PA565041
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATQFTNLQPVA VGHLTTPERQ LQLDNSILTV TFDTVSGSAM LHNRQNDVQI HPQPEASNPQ SVAHFINLTT LV Highest antigen sequence identity to the following orthologs - mouse 95%, rat 96%.

The exact function of PRDM15 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM15
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRDM15
Gene Accession No. P57071
Gene Alias C21orf83; E130018M06Rik; ORF62; PFM12; PFM15; PR domain 15; PR domain containing 15; PR domain zinc finger protein 15; PR domain-containing protein 15; PR/SET domain 15; PRDM15; Zfp298; Zinc finger protein 298; ZNF298
Gene Symbols PRDM15
Host Species Rabbit
Immunogen Recombinant Human PRDM15. Recombinant protein control fragment (Product #RP-105808).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.