Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM16 Monoclonal Antibody (1740CT793.52.44)
GREENER_CHOICE

Catalog No. PIMA537723
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537723 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537723 Supplier Invitrogen™ Supplier No. MA537723
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRDM16 Monoclonal Antibody for Western Blot

PRDM16 is a zinc finger transcription factor and contains an N-terminal PR domain. The reciprocal translocation t(1;3)(p36;q21) occurs in a subset of myelodysplastic syndrome (MDS) and acute myeloid leukemia (AML). This gene is located near the 1p36.3 breakpoint and has been shown to be specifically expressed in the t(1:3)(p36, q21)-positive MDS/AML. The translocation results in the overexpression of a truncated version of this protein that lacks the PR domain, which may play an important role in the pathogenesis of MDS and AML. Recent studies have shown that PRDM16 normally acts as a Smad3 binding protein that may be important for the development of orofacial structures through modulation of the TGF-beta signaling pathway. Other experiments have indicated that PRDM16 controls a bidirectional cell fate switch between skeletal myoblasts and brown fat cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM16
Applications Western Blot
Classification Monoclonal
Clone 1740CT793.52.44
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PRDM16
Gene Accession No. Q9HAZ2
Gene Alias 5730557K01Rik; CMD1LL; csp1; Histone-lysine N-methyltransferase PRDM16; Kiaa1675; line 27; LVNC8; MDS1/EVI1-like gene 1; MEL1; PFM13; PR domain 16; PR domain containing 16; PR domain zinc finger protein 16; PR domain-containing protein 16; Prdm16; Transcription factor MEL1
Gene Symbols PRDM16
Host Species Mouse
Immunogen recombinant protein between 779-996 amino acids from PRDM16.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63976
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.