Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115792 Supplier Invitrogen™ Supplier No. PA5115792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PRDM2.

PRDM2, a member of nuclear/histone methyltransferase superfamily, consists of an N-terminal PR domain, an Rb-binding motif, 8 Kruppel-like zinc finger motifs, Nuclear hormone-receptor binding motif and a C-terminal PR-domain binding motif. Ubiquitously expressed in low levels, PRDM2 exists in 2 isoforms, a 280 kDa RIZ1 containing PR-domain and a 250 kDa truncated RIZ2 that lacks PR-domain. A tumor suppressor with a prominent role in cell growth and differentiation, PRDM2 binds to Retinoblastoma protein, estrogen receptor, GATA-3 and TPA-responsive element (MTE) of Heme-oxygenase-1 gene and influences pathogenesis of Retinoblastoma, transcriptional regulation during neuronal differentiation, transactivation of Heme-oxygenase-1 gene and specific effector of estrogen action. It is inactivated or altered due to mutations and hypermethylation in many types of human cancers suggesting a potential biomarker.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PRDM2
Gene Accession No. Q13029, Q63755
Gene Alias 4833427P12Rik; E330024L24Rik; G3b; GATA-3 binding protein G3B; GATA-3-binding protein G3B; Gm1033; Gm19732; HUMHOXY1; KMT8; lysine N-methyltransferase 8; MTBzf; MTB-ZF; MTE-binding protein; PR domain 2; PR domain containing 2, with ZNF domain; PR domain zinc finger protein 2; PR domain-containing protein 2; Prdm2; retinoblastoma interacting zinc finger protein; retinoblastoma protein-binding zinc finger protein; retinoblastoma protein-interacting zinc finger protein; RIZ; RIZ1; RIZ2; RP5-1177E19.1; zinc finger protein RIZ; zinc-finger DNA-binding protein; Znfpr1c1
Gene Symbols PRDM2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRDM2(Accession Q13029), corresponding to amino acid residues W139-Q189.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110593, 313678, 7799
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.