Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRDM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580930
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PRDM2/KMT8.
PRDM2, a member of nuclear/histone methyltransferase superfamily, consists of an N-terminal PR domain, an Rb-binding motif, 8 Kruppel-like zinc finger motifs, Nuclear hormone-receptor binding motif and a C-terminal PR-domain binding motif. Ubiquitously expressed in low levels, PRDM2 exists in 2 isoforms, a 280 kDa RIZ1 containing PR-domain and a 250 kDa truncated RIZ2 that lacks PR-domain. A tumor suppressor with a prominent role in cell growth and differentiation, PRDM2 binds to Retinoblastoma protein, estrogen receptor, GATA-3 and TPA-responsive element (MTE) of Heme-oxygenase-1 gene and influences pathogenesis of Retinoblastoma, transcriptional regulation during neuronal differentiation, transactivation of Heme-oxygenase-1 gene and specific effector of estrogen action. It is inactivated or altered due to mutations and hypermethylation in many types of human cancers suggesting a potential biomarker.
Specifications
PRDM2 | |
Polyclonal | |
Unconjugated | |
PRDM2 | |
4833427P12Rik; E330024L24Rik; G3b; GATA-3 binding protein G3B; GATA-3-binding protein G3B; Gm1033; Gm19732; HUMHOXY1; KMT8; lysine N-methyltransferase 8; MTBzf; MTB-ZF; MTE-binding protein; PR domain 2; PR domain containing 2, with ZNF domain; PR domain zinc finger protein 2; PR domain-containing protein 2; Prdm2; retinoblastoma interacting zinc finger protein; retinoblastoma protein-binding zinc finger protein; retinoblastoma protein-interacting zinc finger protein; RIZ; RIZ1; RIZ2; RP5-1177E19.1; zinc finger protein RIZ; zinc-finger DNA-binding protein; Znfpr1c1 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
7799 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
Q13029 | |
PRDM2 | |
Recombinant Human PRDM2 protein (Met1-Ala200). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction