Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRDM4 Antibody (RAB-C367), Alexa Fluor™ 750 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320139AF750
Description
PRDM4 Monoclonal antibody specifically detects PRDM4 in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
PRDM4 | |
Monoclonal | |
Alexa Fluor 750 | |
MGC45046, PFM1PR domain-containing protein 4, PR domain containing 4, PR domain zinc finger protein 4, PR-domain zinc-finger protein PFM1 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM4 protein under non-denaturing conditions. | |
0.1 mL | |
Chromatin Research | |
11108 | |
Store at 4°C in the dark. | |
IgG κ |
ELISA, Immunofluorescence | |
RAB-C367 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction