Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM4 Monoclonal Antibody (4B6G1)
GREENER_CHOICE

Catalog No. PIMA531860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531860 Supplier Invitrogen™ Supplier No. MA531860
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31860 has been tested in indirect ELISA.

The protein encoded by this gene is a transcription factor of the PR-domain protein family. It contains a PR-domain and multiple zinc finger motifs. Transcription factors of the PR-domain family are known to be involved in cell differentiation and tumorigenesis. An elevated expression level of this gene has been observed in PC12 cells treated with nerve growth factor, beta polypeptide (NGF). This gene is located in a chromosomal region that is thought to contain tumor suppressor genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM4
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4B6G1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PRDM4
Gene Accession No. Q9UKN5
Gene Alias 1700031E19Rik; 2810470D21Rik; AW552272; MGC45046; PFM1; PR domain 4; PR domain containing 4; PR domain zinc finger protein 4; PR domain-containing protein 4; PRDM4; PR-domain zinc-finger protein PFM1; SC-1
Gene Symbols PRDM4
Host Species Mouse
Immunogen Purified recombinant fragment of human PRDM4 (AA: 476-575) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11108
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.