Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582877 Supplier Invitrogen™ Supplier No. PA582877
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSTSHNLASL ESVSLHEVGL SLEPVAVSSI TQEVAMGTGH VDVSSDSLSF VSPSLQMEDS NSNKENMATL FTIWCTLCDR AYPSDCPEHG PVTFVPDTPI ESRARLSLPK QLVLRQSIVG AEVGVWTGET IPVRTCFGPL IG Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 96%.

The protein encoded by this gene is a transcription factor of the PR-domain protein family. It contains a PR-domain and multiple zinc finger motifs. Transcription factors of the PR-domain family are known to be involved in cell differentiation and tumorigenesis. An elevated expression level of this gene has been observed in PC12 cells treated with nerve growth factor, beta polypeptide (NGF). This gene is located in a chromosomal region that is thought to contain tumor suppressor genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRDM4
Gene Accession No. Q9UKN5
Gene Alias 1700031E19Rik; 2810470D21Rik; AW552272; MGC45046; PFM1; PR domain 4; PR domain containing 4; PR domain zinc finger protein 4; PR domain-containing protein 4; PRDM4; PR-domain zinc-finger protein PFM1; SC-1
Gene Symbols PRDM4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRDM4. Recombinant protein control fragment (Product #RP-89326).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11108
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.