Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRDM5 Antibody (7D4C12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$548.00
Specifications
Antigen | PRDM5 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Classification | Monoclonal |
Description
PRDM5 Monoclonal specifically detects PRDM5 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
PRDM5 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
PFM2PR domain-containing protein 5, PR domain containing 5, PR domain zinc finger protein 5 | |
PRDM5 | |
IgG1 | |
Protein A or G purified |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Chromatin Research, Signal Transduction | |
PBS with 0.05% Sodium Azide | |
11107 | |
Purified recombinant fragment of human PRDM5 (AA: 1-100) expressed in E. Coli. | |
Primary | |
73 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title