Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRDM5 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298837F
Description
PRDM5 Polyclonal antibody specifically detects PRDM5 in Human samples. It is validated for Western BlotSpecifications
PRDM5 | |
Polyclonal | |
Western Blot | |
PFM2PR domain-containing protein 5, PR domain containing 5, PR domain zinc finger protein 5 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PRDM5. | |
0.1 mL | |
Chromatin Research, Signal Transduction | |
11107 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction