Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM5 Monoclonal Antibody (7D4E11)
GREENER_CHOICE

Catalog No. PIMA531862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531862 Supplier Invitrogen™ Supplier No. MA531862
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31862 has been tested in indirect ELISA.

PRDM5, a novel member of PR domain protein family, contains a PR-domain at the -NH2 terminus followed by 16 zinc finger motifs. It is found to have a role in cell differentiation and tumorigenesis. Reports suggest PRDM5 causes G2/M arrest and apoptosis upon infection of tumor cells and has been found to be silenced in breast, ovarian and lung cancers, suggesting a potential clinical significance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM5
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 7D4E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PRDM5
Gene Accession No. Q9NQX1
Gene Alias 4432417F03Rik; 6530401I24Rik; AI197291; BCS2; E130112L17Rik; PFM2; PR domain 5; PR domain containing 5; PR domain zinc finger protein 5; PR domain-containing protein 5; PR/SET domain 5; PRDM5; unm hi61; un-named hi61
Gene Symbols PRDM5
Host Species Mouse
Immunogen Purified recombinant fragment of human PRDM5 (AA: 1-100) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11107
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.