Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543659 Supplier Invitrogen™ Supplier No. PA543659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SHTHTHTHTH THSLTLFCAV VKPTCPAGKL PERGTKEPGR GSGQAQQPTA.

PRDM6 contains 4 C2H2-type zinc fingers and 1 SET domain. PRDM6 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRDM6
Gene Accession No. Q9NQX0
Gene Alias Gm92; PFM3; PR domain 6; PR domain containing 6; PR domain zinc finger protein 6; PR domain-containing protein 6; PR domain-containing protein in smooth muscle; PRDM6; PR-domain zinc finger protein 6; Prism; putative histone-lysine N-methyltransferase PRDM6; RGD1559594
Gene Symbols PRDM6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PRDM6.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93166
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.