Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRDM6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA556439
Description
Immunogen sequence: ELPEWLRDLP REVCLCTSTV PGLAYGICAA QRIQQGTWIG PFQGVLLPPE KVQAGAVRNT QHLWEIYDQD GTLQHFIDGG EPSKSSWMRY IRCARHCGE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.
PRDM6 contains 4 C2H2-type zinc fingers and 1 SET domain. PRDM6 may be involved in transcriptional regulation.
Specifications
PRDM6 | |
Polyclonal | |
Unconjugated | |
PRDM6 | |
PRDM6 | |
Recombit protein corresponding to Human PRDM6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NQX0 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
93166 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction