Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121160
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121160 Supplier Invitrogen™ Supplier No. PA5121160
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, HepG2, BT-474. The target is usually found in the following locations: Chromosome, Nucleus. Immunogen sequence: NFFIDSCAAH GPPTFVKDSA VDKGHPNRSA LSLPPGLRIG PSGIPQAGLG VWNEASDLPL GLHFGPYEGR ITEDEEAANS GYSWLITKGR NCYEYVDGKD K.

This gene encodes a member of a family of proteins that may have roles in transcription and other nuclear processes. The encoded protein contains a KRAB (Kruppel-associated box) domain -A box and a SET (Su(var)3-9, Enhancer-of-zeste, Trithorax) domain and may function as a histone methyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRDM7
Gene Accession No. Q9NQW5
Gene Alias histone-lysine N-methyltransferase PRDM9-like; PFM4; PR domain 7; PR domain containing 7; PR domain containing 9; PR domain zinc finger protein 7; PR domain-containing protein 7; PR/SET domain 7; PRDM7; Prdm9; PR-domain family protein 4; probable histone-lysine N-methyltransferase PRDM7; ZNF910
Gene Symbols PRDM7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 210-310 of human PRDM7 (NP_001091643.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11105
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.