Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRDM9 Antibody (RAB-C370), DyLight 488 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320141G
Description
PRDM9 Monoclonal antibody specifically detects PRDM9 in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
PRDM9 | |
Monoclonal | |
DyLight 488 | |
EC 2.1.1.43, histone-lysine N-methyltransferase PRDM9, minisatellite binding protein 3 (115kD), minisatellite binding protein 3, 115kDa, PFM6MEISETZ, PR domain containing 9, PR domain zinc finger protein 9, PR domain-containing protein 9, PRMD9, ZNF899 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM9 protein under non-denaturing conditions. | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunofluorescence | |
RAB-C370 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
56979 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction